CryoSure® Vials and Caps, Caplugs
Supplier: Caplugs
222-3901-085 222-3902-085 222-3905-085 222-3926-085
76538-730CT 161.4 USD
76538-730 76538-732 76538-734 76538-736
CryoSure® Vials and Caps, Caplugs
Vials
Caplugs Evergreen CryoSure® brand vials are recommended for storing cell cultures, blood/serum specimens, sperm, and other biological specimens at vapor phase* liquid nitrogen temperatures (–320 °F/–195 °C).
  • CryoSure® vials feature external threads designed to be less susceptible to aerosol contamination than internally threaded vials
  • Straight vertical inside walls with no shoulder for a large accessible opening which makes processing and retrieval of samples convenient
  • Ultra-clear polypropylene material - provides superior tensile strength and resistance to gamma radiation sterilization
  • Available in multiple colors
Caplugs Evergreen CryoSure® vial caps fit all sizes of CryoSure® vials. They have a molded-in sealing ring for rim-seal that provides a leakproof closure without the use of a silicone gasket.