Cole-Parmer® Incubators, Natural Convection, Antylia Scientific
Supplier: Antyila Scientific
52411-56 52411-58 52411-60 52411-62 52411-64
76539-142EA 1395.26 USD
76539-142 76539-144 76539-146 76539-148 76539-150
Cole-Parmer® Incubators, Natural Convection, Antylia Scientific
Incubators Standard Incubators
Gravity convection provides a lower chance of sample cross contamination.
  • PID technology for high precision control and small temperature fluctuation
  • Extended-life silicone rubber gasket provides a long service life
  • Features corrosion-resistant stainless steel
  • Anti-skid shelf design, easy to operate by single hand
  • Glass doors ensure perfect observation of the samples
Gravity convection incubators allow heat to fill the chamber naturally, providing a lower chance of cross contamination between samples. These gravity convection incubators have aMore Product Information