1-Octanesulfonic acid sodium salt, anhydrous for HPLC

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 1-octansulfonateSodium octane-1-sulphonate

AC41636-0050
TS41636-0050EA 132 USD
TS41636-0050
1-Octanesulfonic acid sodium salt, anhydrous for HPLC
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://us.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Loss on drying ≤1.5 % (1 g, 110 °C, 75 min)
Solubility (0.1 % in water) Clear and completely mixed
UV (2.7 g in 50 ml water) Acidified to pH 3 with Phosphoric Acid ACS grade
UV at 210 nm A: ≤1.00
UV at 220 nm A: ≤0.10
UV at 254 nm A: ≤0.04
UV at 280 nm A: ≤0.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR