Volumetric Flasks, Class A, Individually Certified, Wide Mouth, Amber Glass, QR, United Scientific Supplies
Supplier: United Scientific Supplies
FGAM005 FGAM010 FGAM020 FGAM025 FGAM050 FGAM100 FGAM200 FGAM250
76531-926EA 45.71 USD
76531-926 76531-928 76531-930 76531-932 76531-934 76531-936 76531-938 76531-940
Volumetric Flasks, Class A, Individually Certified, Wide Mouth, Amber Glass, QR, United Scientific Supplies
Flasks Volumetric Flasks
Borosilicate glass amber-colored to protect light-sensitive materials.
  • Made from borosilicate glass with heavy, uniform wall thickness Durable enamel printing and diamond-etched graduation line
  • Wide mouth allows for easy pipette access
  • Printed QR code enables easy access to calibration certificate using smart device; paper certificate also included
A printed QR code allows for easy access to the calibration certificate from a mobile device or computer. A paper certificate is also included.
Certifications: Complies with ASTM E288 Class A serialized glassware specifications; individually certified and calibrated to contain (TC).