Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence, AAT Bioquest
Supplier: AAT BIOQUEST INC
35999 36000 36001
76483-908EA 249.58 USD
76483-908 76483-910 76483-912
Screen Quest™ Membrane Potential Assay Kit, Orange Fluorescence, AAT Bioquest
Assays Cellular Assays
Membrane potential is the difference in voltage between the interior and exterior of a cell.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.