Metal Fluor™ Zn-520 potassium salt
Supplier: AAT BIOQUEST INC
21262
76483-380EA 240.68 USD
76483-380
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 is designed for detection of higher zinc ion concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists (0.05 to 50 µM) with minimal interfering calcium sensitivity.
  • Cell impermeable
  • Zinc-dependent fluorescence
  • Multiple applications
Caution: For research use only.