iFluor® 647 Goat Anti-Mouse IgG (H+L) *Cross Adsorbed*
Supplier: AAT BIOQUEST INC
16562 16783
76482-510EA 124.31 USD
76482-510 76482-512
iFluor® 647 Goat Anti-Mouse IgG (H+L) *Cross Adsorbed*
Antibodies
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Caution: For research use only.