MagaDye™ 588-ddTTP, AAT Bioquest
Supplier: AAT BIOQUEST INC
17061
17065
76485-276EA
1142.29
USD
76485-276
76485-278
MagaDye™ 588-ddTTP, AAT Bioquest
Nucleic Acid Reagents
Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
- In-house quality control
- Useful for sanger sequencing applications
Caution: For research use only.