Fura-10™ potassium salt
Supplier: AAT BIOQUEST INC
Warning
21110 21111
76485-314EA 117.59 USD
76485-314 76485-316
Fura-10™ potassium salt
Fura-10™ potassium salt
In contrast to single-wavelength indicators such as Fluo-4, the absorption (or fluorescence excitation) maximum of Fura indicators shifts from 380 nm (Fura-2), 415 nm (Fura-8™ and Fura-10™) for the Ca2+-free chelator to about 340 nm (Fura-2), 355 nm (Fura-8™ and Fura-10™) for the Ca2+-bound.
  • Calcium-dependent fluorescence
  • Ratiometric detection
  • Cell impermeant
Caution: For research use only
ADR: 5,II