Fluo-3FF pentapotassium salt

Supplier: AAT BIOQUEST INC
Warning

21019
76483-146EA 361.89 USD
76483-146
Fluo-3FF pentapotassium salt
Fluo-3FF pentapotassium salt

The cell-impermeant Fluo-3FF is an analog of Fluo-3 with much lower calcium binding affinity, Kd ~10 µM.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant


Caution: For research use only

MW: 981,94 g/mol
CAS Number: 348079-14-1
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR