5-Propargylamino-3'-azidomethyl-dCTP
Supplier: AAT BIOQUEST INC
|
Warning
|
17091
76485-654EA
288.22
USD
76485-654
5-Propargylamino-3'-azidomethyl-dCTP
5-Propargylamino-3'-azidomethyl-dCTP
5-Aminopropargyl 3’-azidomethyl dCTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).
- Ngs building block
- Useful for multiple applications
- High quality
Caution: For research use only
Formula:
C13H20N7O13P3 MW: 575,26 g/mol |
CAS Number:
666847-71-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...