Solid and compact manikin for effective training.
- Easy to clean durable, latex free material
- Easy to change disposable airways
- Realistic anatomical landmarks
- Mouth-to-mouth, mouth-to-nose or BVM
- Head-tilt functionality
- Visible chest rise
This high quality and economical AHA compliant BLS manikin is a complete solution to train, monitor, analyze and debrief CPR performances. The manikin connects with the app using a strong and reliable Bluetooth connection – no need to set up a wireless network. The Instructor App provides a detailed performance review of up to six manikins at a time while the Student App enables trainees to directly see and monitor their CPR performance in real-time.
Training Basic Life Support with BASICBilly+ shows how easy you could provide help and save lives through correct cardiac massage and ventilation technique with direct feedback using the apps accordingly. BASICBilly+ has realistic anatomical landmarks to learn correct hand position for chest compression. The head-tilt functionality makes the head easy to tilt in order to open the airways for mouth-to-mouth and mouth-to-nose and BVM resuscitation or simulate obstructed airway. Additionally, the thorax raises as the lungs are ventilated. Mask ventilation is also possible and BASICBilly+ comes with 5 easy to change disposable lung bags and 1 additional face-mask for hygienic artificial ventilation.