VHB™ 3M™ 5952 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-424PK 134.81 USD
76524-424 76524-302 76524-426 76524-304 76524-428 76524-306 76524-308 76524-310 76524-430 76524-312 76524-432 76524-412 76524-414 76524-416 76524-418 76524-420 76524-422 76524-300 76524-446 76524-204 76524-324 76524-206 76524-448 76524-326 76524-208 76524-328 76524-330 76524-450 76524-332 76524-210 76524-452 76524-334 76524-212 76524-454 76524-314 76524-434 76524-316 76524-436 76524-318 76524-438 76524-440 76524-200 76524-442 76524-320 76524-202 76524-444 76524-322 76524-226 76524-468 76524-346 76524-104 76524-228 76524-106 76524-348 76524-108 76524-350 76524-470 76524-110 76524-352 76524-230 76524-472 76524-354 76524-112 76524-232 76524-474 76524-114 76524-356 76524-476 76524-234 76524-336 76524-456 76524-214 76524-338 76524-458 76524-216 76524-218 76524-460 76524-220 76524-462 76524-340 76524-222 76524-464 76524-100 76524-342 76524-466 76524-224 76524-102 76524-344 76524-248 76524-126 76524-368 76524-128 76524-370 76524-490 76524-372 76524-130 76524-492 76524-250 76524-132 76524-374 76524-494 76524-252 76524-376 76524-134 76524-254 76524-496 76524-378 76524-136 76524-256 76524-498 76524-358 76524-116 76524-236 76524-478 76524-118 76524-238 76524-480 76524-482 76524-240 76524-360 76524-484 76524-242 76524-120 76524-362 76524-244 76524-486 76524-364 76524-122 76524-246 76524-488 76524-124 76524-366 76524-028 76524-148 76524-390 76524-392 76524-150 76524-270 76524-394 76524-152 76524-272 76524-030 76524-396 76524-154 76524-032 76524-274 76524-398 76524-156 76524-034 76524-276 76524-158 76524-278 76524-036 76524-138 76524-258 76524-018 76524-260 76524-380 76524-020 76524-262 76524-140 76524-382 76524-022 76524-264 76524-384 76524-142 76524-266 76524-024 76524-144 76524-386 76524-026 76524-268 76524-388 76524-146 76524-170 76524-290 76524-172 76524-292 76524-050 76524-174 76524-294 76524-052 76524-176 76524-296 76524-054 76524-178 76524-056 76524-298 76524-058 76524-038 76524-280 76524-040 76524-282 76524-160 76524-284 76524-042 76524-162 76524-044 76524-286 76524-164 76524-046 76524-288 76524-166 76524-048 76524-168 76524-500 76524-502 76524-504 76524-190 76524-192 76524-070 76524-194 76524-072 76524-196 76524-074 76524-198 76524-076 76524-078 76524-060 76524-180 76524-062 76524-182 76524-064 76524-184 76524-066 76524-186 76524-068 76524-188 76524-402 76524-404 76524-406 76524-408 76524-090 76524-092 76524-094 76524-096 76524-098 76524-410 76524-080 76524-082 76524-084 76524-086 76524-088 76524-400
VHB™ 3M™ 5952 Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • Black, 45 mil thick, (1.1 mm), modified acrylic adhesive and very conformable acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives
  • Maximum temperature: 300 °F (149 °C)
  • Adhesive strength: 22 lbs./inch
VHB™ tape 5952 is very conformable double-sided foam tape. This tape is designed for bonding with medium low surface energy paints, plastics, and powder coated paints. With an extra conformable foam construction, this product offers the maximum performance when adapting to irregular or mismatched bonding surfaces.