Dextran, M.W. 10,000 Cal-520®/zFluor 647™ labelled, RatioWorks™
Supplier: AAT BIOQUEST INC
20604
76485-794EA
579.69
USD
76485-794
Dextran, M.W. 10,000 Cal-520®/zFluor 647™ labelled, RatioWorks™
Dextran
Calcium measurement is critical for numerous biological investigations.
- Well-retained in cells
- 10,000 MW
- Useful for in vivo imaging
- Ratiometric imaging
From Eukaryotic cells
Caution: For research use only.
Formula:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...