Dextran, M.W. 3,000 Cal-590™ labelled
Supplier: AAT BIOQUEST INC
20508
76483-048EA
361.89
USD
76483-048
Dextran, M.W. 3,000 Cal-590™ labelled
Dextran
Calcium measurement is critical for numerous biological investigations.
- Well-retained in neurons
- 3,000 mw
- Useful for in vivo imaging
From Eukaryotic cells
Caution: For research use only.
Formula:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...