MeO-Succ-Arg-Pro-Tyr-AMC
Supplier: AAT BIOQUEST INC
Warning
13460
76482-132EA 118.54 USD
76482-132
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC is a sensitive fluorogenic substrate for chymotrypsin-like proteases.
  • protease substrate
  • bright signal
  • multiple applications
ADR: 5,II