iFluor® 440, Succinimidyl Ester fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1041 71041 71519 71569
76485-640EA 237.74 USD
76485-640 76485-642 76485-644 76485-646
iFluor® 440, Succinimidyl Ester fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR