5-TAMRA fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-CarboxytetramethylrhodamineTAMRA, 5-isomer2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate

365
76480-828EA 2895.22 USD
76480-828
5-TAMRA fluorescent probe
5-TAMRA

5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is widely used to label peptides and proteins. It is preferred for some complicated biological applications where reproducibility is more critical than material cost since the minor positional difference between 5-TAMRA and 6-TAMRA might affect some biological properties of the underlying conjugates. Currently AAT Bioquest is using a proprietary process to synthesize isomer-free 5-TAMRA without the conventional chromatographic purification.


  • High reproducibility

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
MDL Number: MFCD00269769
CAS Number: 91809-66-4
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/33/46/43933346.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR