2',3'-cGAMP, sodium salt second messenger

Supplier: AAT BIOQUEST INC
Warning

20313 20314
76485-120EA 348.69 USD
76485-120 76485-122
2',3'-cGAMP, sodium salt second messenger
2',3'-cGAMP, sodium salt

2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate" adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.


  • Useful for immunity research
  • Multiple applications

CAS Number: 1441190-66-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9dcc8bfc653d406fef1b2ce7078dd915aea942c&vid=b9dcc8bfc653d406fef1b2ce7078dd915aea942c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20faf503aae4376a37873a6a7ca31ec1b6b88272&vid=1b61950c5b2927b8ea1348e330364066b6300f1d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR