Amplite® Fluorimetric Peroxidase (HRP) Assay Kit *Red Fluorescence* , AAT Bioquest
Supplier: AAT BIOQUEST INC
11552 11553
76481-944EA 249.58 USD
76481-944 76481-946
Amplite® Fluorimetric Peroxidase (HRP) Assay Kit *Red Fluorescence* , AAT Bioquest
Assays Enzyme Assays
Peroxidase is a small molecule (MW ~40 KD) that can usually be conjugated to an antibody in a 4:1 ratio.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.