EDGE™ Integrated Electrophoresis System, Edvotek
Supplier: Edvotek
Edge™
500
470336-666EA 299.95 USD
470336-666
EDGE™ Integrated Electrophoresis System, Edvotek
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The EDGE™ Integrated Electrophoresis System revolutionizes biotechnology by merging an electrophoresis chamber, power supply and blue light into a single, safe and easy-to-use apparatus.
  • <10 minute gel runs for fast, real-time results
  • Integrated electrophoresis chamber, power supply and blue light
  • Dual 100/150 Volt modes with automatic safety cutoff switch
  • Includes 10×7 cm gel tray, rubber end caps and (2) 6/8 tooth combs
  • Permanent electrodes - no replacement required
  • Built-in fan and ventilated design prevent condensation
  • Compatible with SYBR® Safe, GelGreen® and other blue light-reactive dyes
  • Ready-to-Run™ EDVOTEK® and PLTW® experiments
  • Universal voltage for worldwide use
  • Three-year warranty
It runs a 10×7 cm gel in less than 10 minutes with crisp band resolution while enabling real-time visualization of DNA migration. And all EDVOTEK® and PLTW® electrophoresis experiments may be run without protocol adjustments. Bring the EDGE™ to your laboratory for the ultimate in convenience, performance and results.