Simple Green d Pro 5 Disinfectant, Essendant
Supplier: Essendant
simple green®
SMP30501CT
76500-278CT 155.02 USD
76500-278
Simple Green d Pro 5 Disinfectant, Essendant
Cleaning Agents Disinfectants
One-step disinfectant and sanitizer. Simple Green d Pro 5 has demonstrated effectiveness against viruses similar to SARS-CoV-2 on hard, non-porous surfaces.
  • Fungicide, mildewstat, virucide, and deodorizer
  • Simple Green d Pro 5 kills Pandemic 2009 H1N1 influenza A virus (formerly called swine flu)
  • For institutional and industrial use
  • Chemical compound: Quaternary Ammonium Chloride
Use Simple Green d Pro 5 in athletic facilities, bars and restaurants, bathrooms, business and office buildings, cafeterias, day care centers, exercise facilities, food preparation areas, hotels and motels, kitchens, nursing homes, pet animal quarters, public restrooms, schools and colleges, USDA inspected food processing facilities, and zoos.