Silicone Tubing for Microsart Membrane Pumps, Pressure-Sided
Supplier: Sartorius
1ZAS0007 1ZAS0007
76500-950EA 35.98 USD
CA76517-962 76500-950
Silicone Tubing for Microsart Membrane Pumps, Pressure-Sided
Tubing Transfer Tubing
Transparent silicone hose is connected to the Microsart® e.jet transfer pump on the pressure side.
The required length depends on the distance between the transfer pump and the spout.
Application: Microbial enumeration
Application area: Water analysis
Industry: Food and beverage; pharma/biotech