Safety Funnels with Cap, SCAT
Supplier: SCAT AMERICAS, INC.
107670 108670 118630
76503-224EA 336.82 USD
76503-224 76503-588 76503-646
Safety Funnels with Cap, SCAT
Funnels Safety Funnels
Safety funnels with hinged lid cap, sieve and safety lance.
  • Lid funnel with double thread R2" BSP/G2" (m) + 2" Tri-Sure (m) or S60/61
  • Made of PE-HD material
  • Stable stainless steel strainer
  • Large funnel diameter: 250 mm
The funnels with hinged lid are suitable for universal use in the laboratory and are highly resistant to conventional chemicals due to the high-quality PE-Hd material.