Brady® B33 Series Polyamide Coated Nylon Cloth Labels, Brady
Supplier: Brady Worldwide
B33-1-499 B33-5-499 B33-88-499 B33-254-499 B33-91-499 B33-87-499 B33-90-499 B33-89-499 B33-325-499 B33-326-499 B33-327-499 B33-328-499
76520-328RL 338.93 USD
76520-328 76520-330 76520-332 76520-334 76520-336 76520-338 76520-340 76520-342 76520-344 76520-346 76520-348 76520-350
Brady® B33 Series Polyamide Coated Nylon Cloth Labels, Brady
Labels
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
  • Recommended ribbon: B30-R6000
  • Low temperature labels with permanent adhesive
  • Permanent acrylic adhesive type
  • Nylon cloth material
  • UL approved, CSA accepted
Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable. Recommended for indoor use. Service temperature of −40 to 193 °F (−40 to 90 °C). A ribbon is required to print this material. The recommended ribbon is B30-R4300.
Order Now
SPECIFICATIONS
Environmentally Preferable