SCIEX 3500 Triple Quad Package
Supplier: SCIEX
5062231
76509-676EA 375580.52 USD
76509-676
SCIEX 3500 Triple Quad Package
Spectrophotometers and Photometers Mass Spectrometers
The Sciex 3500 Triple Quad system offers productivity, reliability, and robustness in a modernized entry-level mass spec for today’s analytical laboratories. The Triple Quad 3500 system offers the speed and precision you want from a modern mass spec system, with the legendary performance and dependability you trust from SCIEX technology.
  • Qurved LINAC® collision cell: More MRMs per injection, space-saving design
  • High-pressure Q0 and QJet® Ion Guide: Enhanced sensitivity through more efficient ion focusing
  • Fast eQ™ Electronics: Compatible with fast LC — achieve wider compound coverage
  • Pulse Counting Detector: High reproducibility and accuracy with the AcQuRate™
  • Turbo V ion source and Curtain Gas interface: Achieve consistent results over long runs of dirty samples
If you're currently running your assays using GC, GC-MS, or HPLC, a bold new world awaits you. Discover how the power of liquid chromatography (LC) coupled with triple quadrupole tandem mass spectrometry (MS/MS) can simplify and consolidate your current assays, dramatically increase your assay quality, and vastly expand the repertoire of assays you can provide – without breaking your budget. With the SCIEX Triple Quad™ 3500 LC-MS/MS system, modern hardware, powerful software, and robust engineering combine to enable lower levels of detection and quantitation than conventional GC and LC workflows, and across a wider range of analytes. You’ll get expert level performance at an entry level price.
Certifications: NRTL certificate and EU Declaration of Conformity.
Delivery information: 3500 Triple Quad package includes enhanced high performance triple quadrupole LC-MS/MS mass spectrometer, Turbo V Ion Source, One standard wet pump kit (roughing pump), Data Acquisition Workstation PC with Win10 64-bit OS, Standard Monitor and Standard parts and labour warranty for one year starting from the completion of instrument commissioning. Includes our StatusScope Remote Monitoring Service and one no-charge Preventative Maintenance (PM) during the one-year warranty period.