J&W VF-Xms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CP8809 CP8806
AGCP8809EA 1409 USD
AGCP8809 AGCP8806
J&W VF-Xms Columns, Agilent Technologies
Chromatography Columns
Provides exceptionally high selectivity for pesticides and delivers high resolution in the shortest analysis time.
  • High arylene modified phase for accurate results
  • Isothermal applications up to 340 °C for a broad application range
  • Ideal for confirmation analysis – a more polar alternative to 5% phenyl columns
  • Ultra-low bleed delivers ultimate sensitivity and signal-to-noise
  • Provides exceptionally high selectivity for semivolatile compounds such as pesticides and delivers high resolution with short analysis time
  • Unique selectivity for chlorinated compounds
  • QC test results for retention index, efficiency, selectivity, and bleed reported with every column
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-Xms is a high arylene modified phase that delivers the ultimate in sensitivity and signal-to-noise ratio, and is the low-bleed, more polar alternative to VF-5ms. Compared to nonpolar 'ms'-type phases, VF-Xms provides exceptionally high selectivity for pesticides and delivers high resolution in the shortest analysis time.