J&W Ultra Inert DB-624 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
121-1324UI 122-1334UI 122-1364UI 123-1334UI 125-1334UI 123-1364UI 123-1334UI-INT 122-1334UI-INT 122-1364UI-INT 123-1364UI-INT 125-1374UI 127-1314UI
AG121-1324UIEA 667 USD
AG121-1324UI AG122-1334UI AG122-1364UI AG123-1334UI AG125-1334UI AG123-1364UI AG1231334UI AG1221334UIINT AG1221364UIINT AG1231364UIINT AG125-1374UI AG127-1314UI
J&W Ultra Inert DB-624 Columns, Agilent Technologies
Chromatography Columns
Designed for trace analysis in environmental, pharmaceutical, and other volatile organic compound (VOC) applications.
  • Environmental volatile organic compounds (VOCs) methods
  • Excellent for US EPA Methods: 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, and 8260
  • Industrial chemical analysis – solvents, petrochemicals, specialty chemicals
  • Food and beverage – alcohols, fusel oils
  • Pharmaceutical residual solvents per USP
  • Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds
  • UI testing ensures premium performance column-to-column
  • Identical selectivity to the industry standard DB-624 – upgrade with no change in method required
  • Optimized by the inventors of DB-624
Agilent J&W DB-624 UI is optimal for the fast analysis of volatile organic compounds (VOCs) in pharmaceutical, environmental, food, and industrial chemical applications. The unique Ultra Inert deactivation process enhances peak shape, improving signal-to-noise levels and increasing sensitivity for qualitative and quantitative analysis.
Ultra inertness processing expands application range with excellent peak shape for low molecular weight acidic compounds. UI testing ensures premium performance column-to-column. Identical selectivity to the industry standard DB-624 – upgrade with no change in method required.