Air Science USA® Trays for Laminar Flow Cabinets, Air Science USA LLC
Supplier: Air Science
TRAY-P5-48-XTS TRAY-P5-24-XTS TRAY-P5-36-XTS
MSPP-TRAYP524XEA 258.75 USD
MSPP-TRAYP524X MSPP-TRAYP548X MSPP-TRAYP536X
Air Science USA® Trays for Laminar Flow Cabinets, Air Science USA LLC
Fume Hoods and Enclosures Laminar Flow Cabinets
Spillage trays for Purair® FLOW cabinets.
  • Black chemical resistant polyproplyene
  • Spill retention lip
  • Easily removeable for cleaning