Air Science® Ductless Fume Hoods, Purair® Basic, Air Science USA LLC
Supplier: Air Science
P5-24-XT-A P5-24-XT-G P5-36-XT-A P5-36-XT-G P5-48-XT-A P5-48-XT-G
MSPP-P524XTAEA 2307.2 USD
MSPP-P524XTA MSPP-P524XTG MSPP-P536XTA MSPP-P536XTG MSPP-P548XTA MSPP-P548XTG
Air Science® Ductless Fume Hoods, Purair® Basic, Air Science USA LLC
Fume Hoods and Enclosures Ductless Fume Hoods
Purair® Basic ductless fume hoods are designed to provide high level performance features at an affordable price.
  • High efficiency EC blower
  • Energy saving LED lighting
  • Filter blockage alarm
  • Protects the operator from fume and (with added HEPA/ULPA filter) particle hazards
Purair® Basic ductless fume hoods are designed to protect the user and the environment from hazardous vapors generated on the work surface.
The Purair® Basic Series ductless fume hoods are compact, ideal for use in laboratory environments where space is limited or where only smallMore Product Information