Stainless Steel Multi-Bucket System, Contec®
Supplier: Contec
2617 2995 2707
33503-124EA 642.4 USD
33503-124 76417-128 76417-130
Stainless Steel Multi-Bucket System, Contec®
Mops
Stainless steel cart with two buckets and downpress wringer.
  • Includes two 27 L (7 gal.) buckets and one cart with four non-marking, autoclavable casters
  • Cart dimensions: 66×61×92 cm (26L×24W×36" H), with handle and casters
  • Buckets, wringer, and cart are manufactured from 304 electropolished stainless steel and are fully autoclavable
  • Buckets are deep drawn and seamless, reducing the risk of microbial contamination
  • A lip around the top edge of the buckets allows for easy lifting
The Stainless steel multi-bucket system offers maximum flexibility and canMore Product Information