Tofacitinib (CP-690550) citrate

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,4R)-4-Methyl-3-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanenitrile citrate, 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanenitrile 2-hydroxypropane-1,2,3-tricarboxylate, Tofacitinib citrate

46653-0010 46653-2500
TS46653-0010EA 577 USD
TS46653-0010 TS46653-2500
Tofacitinib (CP-690550) citrate
Tofacitinib (CP-690550) citrate
Formula: C₁₆H₂₀N₆O·C₆H₈O₇
Melting Pt: 198°C…202°C
Storage Temperature: Refrigerator
MDL Number: MFCD11616529
CAS Number: 540737-29-9

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Solubility (5mg/ml DMSO, warmed) Clear solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR