Tolvaptan

Supplier: Thermo Scientific Chemicals

46535-0010 465352500
TS46535-0010EA 824 USD
TS46535-0010 TS465352500
Tolvaptan
Tolvaptan
Formula: C₂₆H₂₅ClN₂O₃
MW: 448.94 g/mol
Melting Pt: 225.9°C
Storage Temperature: Refrigerator
MDL Number: MFCD09838782
CAS Number: 150683-30-0

Order Now

Specification Test Results

Appearance White to light brown powder
Infrared spectrum Conforms
HPLC ≥ 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR