Nickel etchant TFG semiconductor grade
Supplier: Transene
|
Danger
|
0017000-QT
0017000-GAL
76540-946EA
47
USD
76540-946
76540-962
Nickel etchant TFG semiconductor grade
Nickel etchant
Nickel Etchant TFG is a specially formulated etchant for electroless or electrolytic nickel films. May also be used for nickel vanadium (NiV) films or any nickel film containing non-nickel impurities. Will not attack gallium arsenide (GaAs).
Boiling Pt:
100 °C (water) Density: 1.04 g/cc Storage Temperature: Ambient |
MDL Number:
MFCD09039236 UN: 2796 ADR: 8 II |
Specification Test Results
Appearance | Light Yellow |
Etch Capacity (rate declines at ~70%) | 100 oz/gallon |
Etch Rate at 40°C | 50 Å/sec |
Filtration | 1 um |
Incompatible Materials | Al, Ag |
pH | Strong Acid |
Recommended Operating Temperatures | 20-80°C (30-40°C most common) °C |
Shelf Life | 1 Year |
Storage Conditions | Avoid Freezing |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...