Ammonia solution 29%, Low-Metal (LM) grade semiconductor grade
Supplier: Transene
|
Danger
|
Synonyms:
Ammonia aqueous
, Ammonium hydroxide solution
, Ammonia water
, Ammonium hydroxide
AMMYDLM-QT
AMMYDLM-GAL
76541-060EA
21.29
USD
76541-060
76541-026
Ammonia solution 29%, Low-Metal (LM) grade semiconductor grade
Ammonia solution
Transene Ammonium Hydroxide 29% LM is a "low metals" grade suitable for semiconductor and electronics processing.
Formula:
NH₃ MW: 35.05 g/mol Boiling Pt: 36 °C (water Melting Pt: -72 °C Density: 0.9 g/cc Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 2672 ADR: 8,III |
Specification Test Results
Assay (NH3) | 30 % max. |
Density | 0.91, nominal g/cc |
Color | 10 max. |
Appearance | Clear, sediment-free |
Reducing Substance as KMnO4 | P/F ppm |
Carbon Dioxide (CO2) | 20 ppm max. |
Residue on Ignition | 20 ppm max. |
Non-Volatile Residue | 5 mg/100 mL max. |
Aluminum (Al) | 10 ppb max. |
Barium (Ba) | 10 ppb max. |
Beryllium (Be) | 10 ppb max. |
Bismuth (Bi) | 10 ppb max. |
Boron (B) | 10 ppb max. |
Cadmium (Cd) | 10 ppb max. |
Calcium (Ca) | 10 ppb max. |
Chloride (Cl) | 0.2 ppm max. |
Chromium (Cr) | 10 ppb max. |
Cobalt (Co) | 10 ppb max. |
Copper (Cu) | 10 ppb max. |
Gallium (Ga) | 10 ppb max. |
Germanium (Ge) | 10 ppb max. |
Gold (Au) | 10 ppb max. |
Heavy Metals (as As) | 5 ppb max. |
Iron (Fe) | 10 ppb max. |
Lead (Pb) | 10 ppb max. |
Lithium (Li) | 10 ppb max. |
Magnesium (Mg) | 10 ppb max. |
Manganese (Mn) | 10 ppb max. |
Molybdenum (Mo) | 10 ppb max. |
Nickel (Ni) | 10 ppb max. |
Niobium (Nb) | 10 ppb max. |
Nitrate (NO3) | 0.4 ppm max. |
Phosphate (PO4) | 0.1 ppm max. |
Potassium (K) | 10 ppb max. |
Silicon (Si) | 10 ppb max. |
Silver (Ag) | 10 ppb max. |
Sodium (Na) | 10 ppb max. |
Strontium (Sr) | 10 ppb max. |
Sulfate (SO4) | 0.2 ppm max. |
Tantalum (Ta) | 10 ppb max. |
Thallium (Tl) | 10 ppb max. |
Tin (Sn) | 10 ppb max. |
Titanium (Ti) | 10 ppb max. |
Vanadium (V) | 10 ppb max. |
Zinc (Zn) | 10 ppb max. |
Zirconium (Zr) | 10 ppb max. |
Particle Count, 0.5 um | 150 ct/ml max. |
Particle Count, 1.0 um | 25 ct/ml max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...