Acetic acid ≥99.70%, Low-Metal (LM) grade semiconductor grade
Supplier: Transene
|
Danger
|
Synonyms:
Methanecarboxylic acid
, Vinegar acid
, Ethanoic acid
060-3550-00-GL
060-3550-00-QT
76541-034CS
191.66
USD
76541-034
76541-044
Acetic acid ≥99.70%, Low-Metal (LM) grade semiconductor grade
Acetic acid
Transene Acetic Acid LM grade is a high-purity acetic acid suitable for semiconductor and elecrtonics applications. Low metal ion content is assured. Quart bottle.
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 °C Melting Pt: 16.6 °C Density: 1.05 g/cc Flash Pt: 40 °C |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8,3,II |
Specification Test Results
Acetic Acid | 99.7 % Weight min. |
Acetic Anhydride | 0.01 % Weight max. |
Color | 7 APHA max. |
Chloride | 500 ppb (µmg/g) max. |
Density (at 25 °C) | 1,05 g/mL |
Dilution Test | Pass |
Freezing Point | 61 °F min. |
Heavy Metals | 500 ppb (µmg/g) max. |
Solubility in Water | Pass |
Phosphate | 500 ppb (µmg/g) max. |
Residue after Evaporation | 5000 ppb (µmg/g) max. |
Subst. Reducing Dichromate | Pass |
Subst. Reducing KMnO4 | Pass |
Aluminum | 20 ppb (µmg/g) max. |
Antimony | 5 ppb (µmg/g) max. |
Arsenic | 5 ppb (µmg/g) max. |
Barium | 50 ppb (µmg/g) max. |
Beryllium | 10 ppb (µmg/g) max. |
Bismuth | 50 ppb (µmg/g) max. |
Boron | 20 ppb (µmg/g) max. |
Cadmium | 10 ppb (µmg/g) max. |
Calcium | 100 ppb (µmg/g) max. |
Chromium | 10 ppb (µmg/g) max. |
Cobalt | 10 ppb (µmg/g) max. |
Copper | 20 ppb (µmg/g) max. |
Gallium | 10 ppb (µmg/g) max. |
Germanium | 20 ppb (µmg/g) max. |
Gold | 20 ppb (µmg/g) max. |
Indium | 10 ppb (µmg/g) max. |
Iron | 50 ppb (µmg/g) max. |
Lead | 20 ppb (µmg/g) max. |
Lithium | 10 ppb (µmg/g) max. |
Magnesium | 50 ppb (µmg/g) max. |
Manganese | 10 ppb (µmg/g) max. |
Molybdenum | 20 ppb (µmg/g) max. |
Nickel | 10 ppb (µmg/g) max. |
Platinum | 50 ppb (µmg/g) max. |
Potassium | 50 ppb (µmg/g) max. |
Silver | 10 ppb (µmg/g) max. |
Sodium | 100 ppb (µmg/g) max. |
Strontium | 20 ppb (µmg/g) max. |
Sulfate | 500 ppb (µmg/g) max. |
Thallium | 50 ppb (µmg/g) max. |
Tin | 50 ppb (µmg/g) max. |
Titanium | 50 ppb (µmg/g) max. |
Vanadium | 10 ppb (µmg/g) max. |
Zinc | 20 ppb (µmg/g) max. |
Zirconium | 50 ppb (µmg/g) max. |
Particle >0.5 µm | 150 pcs/mL max. |
Particle >1.0 µm | 10 pcs/mL max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...