High Profile Orbital Shaker, Crystal Industries
Supplier: Crystal
SYC-2102A
490006-550EA 4802.89 USD
490006-550
High Profile Orbital Shaker, Crystal Industries
Shakers and Mixers Orbital Shakers
Orbital shakers can be used for various applications including culturing microbes, washing blots, and general mixing.  The High profile orbital shakers are designed and built for a heavy workload and provide continuous shaking with no vibration or heat output. Crystals orbital shakers can be placed into a standard incubator, converting the incubator into a shaking incubator.
  • Digital control and LED display
  • Low noise level
  • Gentle speed up and slow down
  • Automatic recovery from a power outage
Accessories information: Platform accommodates: 20×25/50 ml, 8×100/250 ml, 6×500 ml, 4×1000 ml, 2×2000/3000 ml, 1×5000 ml flask or 2×small tube racks.
Ordering information: Accessories such as flask clamps, test tube racks (adjustable and non-adjustable), mini universal spring platforms, fixtures for separating funnel, infusion bottle clamps, 96-well microplate holder, and sticky pads are available and must be ordered separately.
Delivery information: Supplied with standard platform and non-skid mat.