Acridine Orange 10-nonyl bromide (NAO) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Nonyl acridine orange3,6-Bis(dimethylamino)-10-nonylacridinium bromideNAO10-Nonylacridine orange bromide

22205
76483-564EA 124.31 USD
76483-564
Acridine Orange 10-nonyl bromide (NAO) ≥95%
Acridine Orange 10-nonyl bromide (NAO)

10-n-Nonyl acridine orange (NAO) is a mitochondrial-specific fluorochrome.

Formula: C₂₆H₃₈BrN₃
MW: 472.50 g/mol
MDL Number: MFCD00467605
CAS Number: 75168-11-5
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR