Indo-1 AM ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
21030
21033
76483-166EA
124.31
USD
76483-166
76483-168
Indo-1 AM ≥95%
Indo-1 AM
Calcium measurements are critical for numerous biological investigations.
Formula:
C₄₇H₅₁N₃O₂₂ |
MDL Number:
MFCD00036955 CAS Number: 112926-02-0 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...