Propidium Iodide ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
17515
17516
76482-858EA
124.31
USD
76482-858
76482-860
Propidium Iodide ≥95%
Propidium iodide
Propidium iodide (PI) belongs to the same chemical class of ethidium bromide.
Formula:
C₂₇H₃₄I₂N₄ MW: 668.4 g/mol |
MDL Number:
MFCD00011921 CAS Number: 25535-16-4 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...