5-Bromo-2'-deoxyuridine (broxuridine) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-Bromo-2'-deoxyuridine5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

17030
76482-824EA 93.54 USD
76482-824
5-Bromo-2'-deoxyuridine (broxuridine) ≥95%
5-Bromo-2'-deoxyuridine (Broxuridine)

5-Bromo-2-deoxyuridine (BrdU) is an analog of thymidine, and can be incorporated into the newly synthesized DNA of replicating cells (S-phase cells).

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
MDL Number: MFCD00006529
CAS Number: 59-14-3
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR