D-(-)-Luciferin potassium salt ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
Beetle Luciferin potassium salt
, Luciferin potassium salt
, Firefly luciferin potassium salt
, (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt
, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
12505
12506
12507
76481-998EA
300.63
USD
76481-998
76481-996
76481-994
D-(-)-Luciferin potassium salt ≥95%
D(-)-Luciferin potassium salt
Luciferin is the most popular and versatile bioluminescent substrate.
Formula:
C₁₁H₇KN₂O₃S₂ MW: 318.41 g/mol |
MDL Number:
MFCD00044928 CAS Number: 115144-35-9 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...