Luminol (3-Aminophthalhydrazide) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-Amino-2,3-dihydrophthalazine-1,4-dioneNSC 50645-Amino-2,3-Dihydro-1,4-Phthalazinedione

11050
76481-892EA 93.54 USD
76481-892
Luminol (3-Aminophthalhydrazide) ≥95%
Luminol (3-Aminophthalhydrazide)

Luminol [3-Aminophthalhydrazide] is a luminescent peroxidase substrate.

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD00006890
CAS Number: 521-31-3
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR