Biocytin ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: ε-Biotinoyl-L-lysineH-Lys(biotinyl)-OH

3080
76481-648EA 180.84 USD
76481-648
Biocytin ≥90%
Biocytin

Biocytin is invaluable as a rapidly transporting, sensitive neuronal tracer requiring little permeabilizing agents.

Formula: C₁₆H₂₈N₄O₄S
MW: 372.484 g/mol
MDL Number: MFCD00077319
CAS Number: 576-19-2
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR