Trypan blue ≥75%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium saltCI 23850Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate)Direct Blue 14AzidinBlueSodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium saltBenzamine blueTrypan blue sodium salt3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium saltDiphenyl Blue

2450
76481-554EA 1195.79 USD
76481-554
Trypan blue ≥75%
Trypan Blue

Trypan blue, a diazo dye is a vital stain that colors dead tissues or cells blue.

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
MDL Number: MFCD00003969
CAS Number: 72-57-1
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR