5-ROX ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-ROX, 5-Carboxy-X-rhodamine

381 385
76480-856EA 4721.06 USD
76480-856 76480-858
5-ROX ≥95%
5-ROX

5-ROX is the purified single isomer of 5(6)-ROX.

Formula: C₃₃H₃₀N₂O₅
MDL Number: MFCD28355897
CAS Number: 216699-35-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6234d68ecd61ce35c6414f160f819db1d4b8af8c&vid=279766db37d78d43ba9d26ff8a3f5d12dbba92c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR