5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-Carboxytetramethylrhodamine N-succinimidyl ester5-TAMRA, SE5-TAMRA-N-succinimidyl esterTAMRA SE, 5 isomer

375 374 373
76480-842EA 129.69 USD
76480-842 76480-844 76480-846
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥95%
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE)

The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
CAS Number: 150810-68-7
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/33/49/43933349.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR