5-TAMRA ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5-CarboxytetramethylrhodamineTAMRA, 5-isomer2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate

363 364
76480-824EA 93.54 USD
76480-824 76480-826
5-TAMRA ≥95%
5-TAMRA

5-TAMRA is the purified single isomer of 5(6)-TAMRA.

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
MDL Number: MFCD00269769
CAS Number: 91809-66-4
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR