5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5(6)-FAM SENHS-FluoresceinFLUOS

112 110 111
76480-562EA 249.58 USD
76480-562 76480-564 76480-560
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥95%
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.

Formula: C₂₅H₁₅NO₉
MW: 473.39 g/mol
MDL Number: MFCD04119713
CAS Number: 117548-22-8
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/33/24/43933324.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR