5(6)-Carboxyfluorescein ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid

100 101 102
76480-542EA 124.31 USD
76480-542 76480-544 76480-546
5(6)-Carboxyfluorescein ≥95%
5(6)-Carboxyfluorescein

Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.

Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
MDL Number: MFCD00151081
CAS Number: 72088-94-9
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c266e82127bb9214ac06e1da48806d25cdeaa4a&vid=16b52fecf5798aaa1efb60ac1f30d13a87cdad70&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR